DL-Valine methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-Val-OMe.HCl, H-DL-Val-OMe·HCl

A318140-100G A318140-10G A318140-25G A318140-5G A318140-500G
103836-092EA 13 USD
103836-092 76978-026 76978-028 77306-168 77306-170 77672-632
DL-Valine methyl ester hydrochloride 97%
DL-Valine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 167.63 g/mol
Melting Pt: 120…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00066118
CAS Number: 5619-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR