DL-tert-Leucine (H-DL-Tle-OH) 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-Tle-OH

A284557-25G A284557-5G A284557-100G A284557-1G A284557-10G
103813-642EA 5 USD
103813-642 103834-774 76837-300 76837-302 77153-888 77305-462 77685-808
DL-tert-Leucine (H-DL-Tle-OH) 97%
DL-tert-Leucine (H-DL-Tle-OH)

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.2 g/mol
Flash Pt: 85.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00065933
CAS Number: 33105-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR