Methyl 2-amino-2-phenylacetate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-Phg-OMe·HCl, H-DL-Phg-Ome.HCl

A140043-100G A140043-10G A140043-1G A140043-25G A140043-5G
103802-818EA 5 USD
103802-818 76972-552 76972-554 77315-926 77315-928 77315-930
Methyl 2-amino-2-phenylacetate hydrochloride 98%
Methyl 2-amino-2-phenylacetate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₂
MW: 201.65 g/mol
Melting Pt: 220…223 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01436195
CAS Number: 15028-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR