H-DL-Phe-OtBu·HCl 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-DL-Phe-OtBu.HCl
A231901-100G
A231901-10G
A231901-1G
A231901-25G
A231901-5G
103810-802EA
10
USD
103810-802
103850-556
76801-104
76801-106
76801-108
77089-462
77089-464
77089-466
H-DL-Phe-OtBu·HCl 95%
H-DL-Phe-OtBu·HCl
Precaución: for Research Use Only
Formula:
C₁₃H₁₉NO₂·HCl MW: 257.8 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00190980 CAS Number: 75898-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...