DL-Phenylalanine methyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-Phe-OMe·HCl, Methyl 2-amino-3-phenylpropanoate hydrochloride

A124561-100G A124561-10G A124561-25G A124561-5G A124561-1000G A124561-500G
76766-822EA 40 USD
76766-822 76766-824 76766-826 77298-004 77876-900 77876-901
DL-Phenylalanine methyl ester hydrochloride 98%
DL-Phenylalanine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₂
MW: 215.677 g/mol
Melting Pt: 158…162 °C
Flash Pt: 126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066113
CAS Number: 5619-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR