4-Nitro-DL-phenylalanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-DL-Phe(4-NO₂)-OH, H-p-Nitro-DL-Phe-OH, H-DL-Phe(4-NO2)-OH
A538804-25G
A538804-5G
A538804-100G
A538804-10G
103839-738EA
11
USD
103839-738
76843-782
76843-784
77253-652
77253-654
4-Nitro-DL-phenylalanine 97%
4-Nitro-DL-phenylalanine
Precaución: for Research Use Only
Formula:
C₉H₁₀N₂O₄ MW: 210.189 g/mol Boiling Pt: 414.1 °C (760 mm Hg) Melting Pt: 236…237 °C Flash Pt: 204.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007384 CAS Number: 2922-40-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...