2-Amino-3-(3-bromophenyl)propanoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-DL-phenylalanine, H-DL-Phe(3-Br)-OH
A183566-100G
A183566-100MG
A183566-10G
A183566-250MG
A183566-25G
A183566-5G
A183566-1G
A183566-500G
103815-646EA
8
USD
103815-646
103846-208
76974-524
76974-526
76974-528
76974-530
77083-814
77083-816
77301-836
77671-592
2-Amino-3-(3-bromophenyl)propanoic acid 98%
2-Amino-3-(3-bromophenyl)propanoic acid
Precaución: for Research Use Only
Formula:
C₉H₁₀BrNO₂ MW: 244.088 g/mol Boiling Pt: 368.4 °C (760 mm Hg) Melting Pt: 226…231 °C Flash Pt: 176.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD05227935 CAS Number: 30163-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...