Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 98%

Supplier: Ambeed
Warning

Synonyms: H-Gly-OBzl·HCl, H-DL-Gly-OBzl hydrochloride, Benzyl glycinate hydrochloride, H-DL-Gly-OBzl.HCl

A323496-100G A323496-10G A323496-1G A323496-25G A323496-5G A323496-500G
103836-282EA 6 USD
103836-282 77251-188 77251-190 77251-192 77306-300 77306-302 77746-629
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 98%
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl)

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₂
MW: 201.65 g/mol
Boiling Pt: 245.5 °C (760 mm Hg)
Melting Pt: 138…140 °C
Flash Pt: 110.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00035442
CAS Number: 2462-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR