H-D-Leu-OtBu HCl 95%

Supplier: Ambeed
Warning

Synonyms: H-D-Leu-OtBu·HCl, (R)-tert-Butyl 2-amino-4-methylpentanoate hydrochloride

A189146-1G A189146-25G A189146-5G A189146-100G A189146-10G
103816-162EA 6 USD
103816-162 103846-754 77084-508 77084-510 77084-512 77685-446 77685-447
H-D-Leu-OtBu HCl 95%
H-D-Leu-OtBu HCl

Precaución: for Research Use Only

Formula: C₁₀H₂₂ClNO₂
MW: 223.7402 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00083642
CAS Number: 13081-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR