DL-Alanine ethyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-aminopropanoate hydrochloride, H-DL-Ala-OEt.HCl

A138523-5G A138523-25G A138523-100G A138523-500G
77298-996EA 12 USD
77298-996 77315-658 77747-981 77747-982
DL-Alanine ethyl ester hydrochloride 98%
DL-Alanine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO₂
MW: 153.606 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013018
CAS Number: 617-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR