(R)-Piperidine-2-carboxylic acid methyl ester hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: H-D-HoPro-OMe.HCl, H-D-Homopro-OMe·HCl

A179627-1G A179627-250MG
103807-446EA 8 USD
103807-446 103845-798 77083-266 77111-366
(R)-Piperidine-2-carboxylic acid methyl ester hydrochloride 95%
(R)-Piperidine-2-carboxylic acid methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
MW: 179.644 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237412
CAS Number: 18650-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR