(R)-Piperidine-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Homopro-OH, D(+)-Pipecolinic acid, (R)-(+)-2-Piperidinecarboxylic Acid, H-D-HoPro-OH, D-Pipecolinic Acid, D-Homoproline
A257502-1G
A257502-5G
A257502-100G
A257502-10G
A257502-500G
103812-308EA
5
USD
103812-308
103852-176
77091-682
77091-684
77166-118
77166-120
77166-122
(R)-Piperidine-2-carboxylic acid 97%
(R)-Piperidine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Melting Pt: 272 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064346 CAS Number: 1723-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...