D-(+)-Homoserine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Hser-OH, H-D-HomoSer-OH, H-D-Homoser-OH, (R)-(+)-Homoserine, (R)-2-Amino-4-hydroxybutyric acid, D-Homoserine
A237134-100G
A237134-10G
A237134-250MG
A237134-1G
A237134-5G
A237134-25G
103811-132EA
7
USD
103811-132
103850-908
76826-638
76826-640
76826-642
77089-976
77089-978
77304-072
D-(+)-Homoserine 97%
D-(+)-Homoserine
Precaución: for Research Use Only
Formula:
C₄H₉NO₃ Boiling Pt: 368.7 °C (760 mm Hg) Melting Pt: 205 °C Flash Pt: 176.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077786 CAS Number: 6027-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...