H-D-Glu(OtBu)-OMe·HCl 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Glu(OtBu)-OMe.HCl, (R)-5-tert-Butyl 1-methyl 2-aminopentanedioate hydrochloride
A301861-10G
A301861-1G
A301861-25G
A301861-5G
103814-312EA
16
USD
103814-312
103835-466
76827-878
77305-830
77305-832
77305-834
H-D-Glu(OtBu)-OMe·HCl 97%
H-D-Glu(OtBu)-OMe·HCl
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₄·HCl MW: 253.8 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973242 CAS Number: 16948-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...