Dimethyl-D-glutamate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Glu(OMe)-OMe.HCl, H-D-Glu(OMe)-OMe·HCl, D-Glutamic Acid Dimethyl Ester Hydrochloride
A144542-1G
A144542-25G
A144542-5G
103803-416EA
5
USD
103803-416
103841-492
77299-396
77316-740
77316-742
Dimethyl-D-glutamate hydrochloride 97%
Dimethyl-D-glutamate hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₃NO₄ MW: 211.64 g/mol Melting Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00235872 CAS Number: 27025-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...