H-D-Glu(OMe)-OH 97%

Supplier: Ambeed
Warning

A223005-100G A223005-10G A223005-250MG A223005-1G A223005-25G A223005-5G
103819-016EA 7 USD
103819-016 103849-894 76770-906 76770-908 76770-910 77088-508 77088-510 77088-512
H-D-Glu(OMe)-OH 97%
H-D-Glu(OMe)-OH

Precaución: for Research Use Only

Formula: C₆H₁₁NO₄
Boiling Pt: 316.1 °C (760 mm Hg)
Melting Pt: 175…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237069
CAS Number: 6461-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR