H-D-Glu(OEt)-OEt.HCl 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Glu(OEt)-OEt·HCl
A175025-25G
A175025-5G
A175025-1G
103845-296EA
11
USD
103845-296
77082-610
77082-612
77096-606
H-D-Glu(OEt)-OEt.HCl 95%
H-D-Glu(OEt)-OEt.HCl
Precaución: for Research Use Only
Formula:
C₉H₁₈ClNO₄ MW: 239.7 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11111544 CAS Number: 1001-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...