H-D-Dap(Boc)-OMe 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Dap(Boc)-OMe·HCl

A159875-250MG A159875-1G A159875-5G
103805-300EA 158 USD
103805-300 103843-526 77110-616 77300-426 77300-428
H-D-Dap(Boc)-OMe 97%
H-D-Dap(Boc)-OMe

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₄
MW: 218.253 g/mol
Boiling Pt: 325.7 °C (760 mm Hg)
Flash Pt: 150.8 °C
Storage Temperature: Freezer
MDL Number: MFCD05663450
CAS Number: 363191-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR