S-Trityl-D-cysteine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-Amino-3-(tritylthio)propanoic acid, H-D-Cys(Trt)-OH
A168965-250MG
A168965-5G
A168965-1G
A168965-100G
A168965-10G
103806-338EA
14
USD
103806-338
103844-594
76784-144
77081-664
77300-954
77667-358
77667-360
S-Trityl-D-cysteine 97%
S-Trityl-D-cysteine
Precaución: for Research Use Only
Formula:
C₂₂H₂₁NO₂S Boiling Pt: 524.7 °C (760 mm Hg) Flash Pt: 271.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00236948 CAS Number: 25840-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...