cis-4-Hydroxy-D(+)-proline 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline

A180127-25G A180127-5G A180127-100G A180127-10G A180127-1G
103807-512EA 5 USD
103807-512 103845-866 77083-348 77083-350 77287-162 77287-164 77301-664
cis-4-Hydroxy-D(+)-proline 98%
cis-4-Hydroxy-D(+)-proline

Precaución: for Research Use Only

Formula: C₅H₉NO₃
MW: 131.13 g/mol
Boiling Pt: 355.2 °C (760 mm Hg)
Melting Pt: 243 °C
Flash Pt: 168.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00005252
CAS Number: 2584-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR