D-2-Cyclohexylglycine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-alpha-Aminocyclohexylacetic Acid, H-D-Chg-OH

A153267-100G A153267-10G A153267-1G A153267-25G A153267-5G
103804-482EA 5 USD
103804-482 103842-654 77190-870 77190-872 77318-298 77318-300 77318-302
D-2-Cyclohexylglycine 97%
D-2-Cyclohexylglycine

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 292.8 °C (760 mm Hg)
Melting Pt: 256 °C
Flash Pt: 130.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01311678
CAS Number: 14328-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR