D-(-)-Aspartic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
A143992-1000G
A143992-100G
A143992-500G
A143992-25G
103841-432EA
5
USD
103841-432
77010-474
77010-476
77010-478
77316-670
D-(-)-Aspartic acid 98%
D-(-)-Aspartic acid
Precaución: for Research Use Only
Formula:
C₄H₇NO₄ Boiling Pt: 283 °C (760 mm Hg) Melting Pt: 124…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063081 CAS Number: 1783-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...