(R)-Dimethyl-2-aminosuccinate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Asp(OMe)-OMe.HCl, D-Aspartic acid dimethyl ester hydrochloride, H-D-Asp(OMe)-OMe·HCl

A229112-100G A229112-10G A229112-25G A229112-5G
103850-350EA 5 USD
103850-350 76815-114 76815-116 77089-170 77089-172
(R)-Dimethyl-2-aminosuccinate hydrochloride 97%
(R)-Dimethyl-2-aminosuccinate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₄
MW: 197.615 g/mol
Melting Pt: 114…119 °C
Storage Temperature: Freezer
MDL Number: MFCD00070384
CAS Number: 69630-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR