H-L-Dap(boc)-OMe·HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-Dapa(Boc)-OMe.HCl, H-Dap(Boc)-OMe·HCl

A213709-250MG A213709-1G A213709-25G
103818-316EA 68 USD
103818-316 103849-024 76814-654 77087-534 77087-536
H-L-Dap(boc)-OMe·HCl 97%
H-L-Dap(boc)-OMe·HCl

Precaución: for Research Use Only

Formula: C₉H₁₉ClN₂O₄
MW: 254.711 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798684
CAS Number: 114559-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR