(R)-tert-Butyl-2-aminopropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: D-Alanine tert-butyl ester hydrochloride, tert-Butyl (R)-2-Aminopropionate Hydrochloride, (R)-2-Aminopropionic Acid tert-Butyl Ester Hydrochloride, H-D-Ala-OtBu.HCl, H-D-Ala-OtBu·HCl

A124410-10G A124410-25G A124410-1G A124410-100G
103800-526EA 6 USD
103800-526 77108-752 77108-754 77312-662 77571-466
(R)-tert-Butyl-2-aminopropanoate hydrochloride 97%
(R)-tert-Butyl-2-aminopropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂·HCl
MW: 181.7 g/mol
Melting Pt: 170…175 °C
Flash Pt: 78.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00077087
CAS Number: 59531-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR