D-Alanine methyl ester hydrochloride 90%
Supplier: Ambeed
|
Warning
|
Synonyms:
D-Alanine Methyl Ester Hydrochloride, (R)-2-Aminopropionic acid Methyl Ester Hydrochloride, H-D-Ala-OMe.HCl, H-D-Ala-OMe·HCl, Methyl D-alaninate hydrochloride, Methyl (R)-2-Aminopropionate Hydrochloride, (R)-2-Aminopropionic Acid Methyl Ester Hydrochloride, (R)-Methyl 2-aminopropionate hydrochloride
A259250-100G
A259250-10G
A259250-500G
A259250-25G
A259250-5G
A259250-1000G
103852-306EA
6
USD
103852-306
76827-098
76827-100
76827-102
77091-848
77091-850
77612-482
D-Alanine methyl ester hydrochloride 90%
D-Alanine methyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₉NO₂ MW: 139.58 g/mol Melting Pt: 108…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066141 CAS Number: 14316-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...