(R)-2-Aminopropanamide hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Ala-NH2.HCl, H-D-Ala-NH₂·HCl, D-Alaninamide hydrochloride
A202883-100G
A202883-10G
A202883-25G
A202883-5G
A202883-1G
A202883-500G
103817-420EA
5
USD
103817-420
103848-068
76814-334
76814-336
77086-184
77086-186
77302-834
77883-467
(R)-2-Aminopropanamide hydrochloride 97%
(R)-2-Aminopropanamide hydrochloride
Precaución: for Research Use Only
Formula:
C₃H₉ClN₂O MW: 124.57 g/mol Boiling Pt: 247.4 °C (760 mm Hg) Melting Pt: 169 °C Flash Pt: 103.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039093 CAS Number: 71810-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...