(R)-(-)-2-Aminobutyric acid 97%

Supplier: Ambeed
Warning

A169665-100G A169665-500G A169665-25G A169665-5G A169665-10G
103844-658EA 5 USD
103844-658 76998-170 76998-172 77081-746 77300-990 77671-402
(R)-(-)-2-Aminobutyric acid 97%
(R)-(-)-2-Aminobutyric acid

Precaución: for Research Use Only

Formula: C₄H₉NO₂
Melting Pt: 308…310 °C
Storage Temperature: Ambient
MDL Number: MFCD00064414
CAS Number: 2623-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR