(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
L-Cysteine ethyl ester hydrochloride, H-Cys-Oet.HCl, H-Cys-OEt·HCl
A253365-100G
A253365-10G
A253365-500G
A253365-25G
A253365-5G
103851-942EA
5
USD
103851-942
76874-670
76874-672
76874-674
77304-612
77304-614
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride 97%
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₁NO₂S·HCl MW: 185.666 g/mol Boiling Pt: 205.9 °C (760 mm Hg) Melting Pt: 123…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012631 CAS Number: 868-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...