S-Trityl-L-cysteine 95%

Supplier: Ambeed
Warning

Synonyms: H-Cys(Trt)-OH

A106724-100G A106724-100MG A106724-10G A106724-500G
103822-820EA 7 USD
103822-820 77107-698 77107-700 77107-702 77107-704
S-Trityl-L-cysteine 95%
S-Trityl-L-cysteine

Precaución: for Research Use Only

Formula: C₂₂H₂₁NO₂S
MW: 363.5 g/mol
Boiling Pt: 524.7 °C (760 mm Hg)
Melting Pt: 182…183 °C
Flash Pt: 271.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00002611
CAS Number: 2799-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR