L-Cyclopropylglycine 97%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Cyclopropyl-L-glycine, (S)-2-Amino-2-cyclopropylacetic acid, H-Cyclopropyl-Gly-OH
A111796-10G
A111796-250MG
A111796-1G
A111796-100G
A111796-25G
103797-904EA
41
USD
103797-904
103823-726
77160-784
77160-786
77284-182
77570-664
77683-024
L-Cyclopropylglycine 97%, Ambeed.Inc
L-Cyclopropylglycine
Precaución: For research use only.
Formula:
C₅H₉NO₂ MW: 115.132 g/mol Boiling Pt: 253.5 °C (760 mm Hg) Flash Pt: 104.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659116 CAS Number: 49606-99-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...