(S)-2-Amino-2-cyclopentylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: H-Cyclopentyl-Gly-OH, L-Cyclopentylglycine

A129949-100MG A129949-10G A129949-250MG A129949-1G A129949-25G
103801-354EA 32 USD
103801-354 76996-310 76996-312 76996-314 77313-764 77313-766
(S)-2-Amino-2-cyclopentylacetic acid 97%
(S)-2-Amino-2-cyclopentylacetic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.18 g/mol
Boiling Pt: 276.6 °C (760 mm Hg)
Flash Pt: 119.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03788076
CAS Number: 2521-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR