trans-4-Cyclohexyl-L-proline 97%

Supplier: Ambeed
Warning

Synonyms: (2S,4S)-4-Cyclohexylpyrrolidine-2-carboxylic acid

A139832-10G A139832-1G A139832-25G A139832-5G
103802-780EA 11 USD
103802-780 76798-182 77315-872 77315-874 77315-876
trans-4-Cyclohexyl-L-proline 97%
trans-4-Cyclohexyl-L-proline

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₂
MW: 197.27 g/mol
Boiling Pt: 343.9±35.0 °C (760 mm Hg)
Melting Pt: 260…262 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08061929
CAS Number: 103201-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR