(S)-2-Amino-2-cyclohexylacetic acid 97%

Supplier: Ambeed
Warning

A162809-1000G A162809-100G A162809-10G A162809-500G A162809-25G A162809-5G
103843-910EA 5 USD
103843-910 77191-144 77191-146 77191-148 77191-150 77319-982 77319-984
(S)-2-Amino-2-cyclohexylacetic acid 97%
(S)-2-Amino-2-cyclohexylacetic acid

Precaución: Sealed in dry, at room temperature.

Formula: C₈H₁₅NO₂
Boiling Pt: 292.8 °C (760 mm Hg)
Melting Pt: 256 °C
Storage Temperature: Ambient
MDL Number: MFCD01311679
CAS Number: 14328-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR