L-4-Benzoylphenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(4-benzoylphenyl)propanoic acid, H-Bpa-OH, H-p-Bz-Phe-OH, H-Phe(4-Bz)-OH

A126709-250MG A126709-25G A126709-5G A126709-10G
103800-852EA 22 USD
103800-852 76797-690 77313-082 77313-084 77566-456
L-4-Benzoylphenylalanine 98%
L-4-Benzoylphenylalanine

Precaución: For research use only.

Formula: C₁₆H₁₅NO₃
MW: 269.3 g/mol
Boiling Pt: 467.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00238102
CAS Number: 104504-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR