1-Methyl-L-aspartate 96%

Supplier: Ambeed
Warning

Synonyms: H-Asp-OMe, (S)-3-Amino-4-methoxy-4-oxobutanoic acid, L-Aspartic acid 1-methyl ester

A120595-250MG A120595-1G A120595-5G A120595-25G A120595-100G
103799-964EA 8 USD
103799-964 76995-790 77297-776 77312-070 77566-232 77599-734
1-Methyl-L-aspartate 96%
1-Methyl-L-aspartate

Precaución: For research use only.

Formula: C₅H₉NO₄
MW: 147.13 g/mol
Boiling Pt: 272.7 °C (760 mm Hg)
Melting Pt: 186 °C
Flash Pt: 118.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00080289
CAS Number: 17812-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR