Di-tert-butyl L-aspartate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Asp(OtBu)-OtBu.HCl, H-Asp(OtBu)-OtBu·HCl, L-Aspartic Acid Di-tert-butyl Ester Hydrochloride, (S)-Di-tert-butyl 2-aminosuccinate hydrochloride

A167075-100G A167075-1G A167075-10G A167075-500G
103806-126EA 5 USD
103806-126 103844-388 76998-056 77320-672 77600-340 77671-362
Di-tert-butyl L-aspartate hydrochloride 98%
Di-tert-butyl L-aspartate hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₄
Flash Pt: 159.3 °C
Storage Temperature: Freezer
MDL Number: MFCD00034851
CAS Number: 1791-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR