H-Asn-OtBu 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2,4-diamino-4-oxobutanoate, H-Asn-OtBu, L-Asparagine tert-butyl ester

A120300-100MG A120300-10G A120300-250MG A120300-25G A120300-1G A120300-5G
103799-914EA 16 USD
103799-914 76995-762 76995-764 76995-766 76995-768 77297-750 77312-022
H-Asn-OtBu 95%
H-Asn-OtBu

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₃
Boiling Pt: 340.2 °C (760 mm Hg)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00190755
CAS Number: 25456-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR