H-Asn(Trt)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Nγ-Trityl-L-Asparagine, Ngamma-Trityl-L-asparagine, (S)-2-Amino-4-oxo-4-(tritylamino)butyric acid

A266087-100G A266087-10G A266087-25G A266087-1G
103812-780EA 9 USD
103812-780 103852-680 77001-034 77001-036 77092-378 77304-946
H-Asn(Trt)-OH 95%
H-Asn(Trt)-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₂N₂O₃
MW: 374.430 g/mol
Boiling Pt: 641.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00190757
CAS Number: 132388-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR