L-Arginine methyl ester dihydrochloride 98%

Supplier: Ambeed
Warning

A128247-10G A128247-1G A128247-500G A128247-1000G A128247-5G A128247-25G A128247-100G
103808-372EA 371 USD
103808-372 77109-032 77109-034 77109-036 77298-276 77298-278 77313-394 77611-122
L-Arginine methyl ester dihydrochloride 98%
L-Arginine methyl ester dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆N₄O₂
Boiling Pt: 329.9 °C (760 mm Hg)
Melting Pt: 190 °C
Flash Pt: 153.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00038948
CAS Number: 26340-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR