H-Arg(pbf)-OH 97%

Supplier: Ambeed
Warning

Synonyms: H-Arg(Pbf)-OH

A176581-100G A176581-100MG A176581-10G A176581-250MG A176581-5G A176581-1G
103807-152EA 8 USD
103807-152 103845-480 76998-382 76998-384 76998-386 76998-388 77082-830 77301-454
H-Arg(pbf)-OH 97%
H-Arg(pbf)-OH

Precaución: for Research Use Only

Formula: C₁₉H₃₀N₄O₅S
Boiling Pt: 636.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00236797
CAS Number: 200115-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR