L-Alanine-tert-butyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: L-Alanine tert-Butyl Ester Hydrochloride, tert-Butyl (S)-2-Aminopropionate Hydrochloride, H-Ala-OtBu·HCl, (S)-2-Aminopropionic acid tert-Butyl Ester Hydrochloride, H-Ala-OtBu.HCl, (S)-2-Aminopropionic Acid tert-Butyl Ester Hydrochloride

A133531-100G A133531-10G A133531-1G A133531-500G A133531-25G A133531-5G
77272-102EA 80.34 USD
77272-102 77272-104 77272-106 77272-108 77314-544 77314-546
L-Alanine-tert-butyl ester hydrochloride 98%
L-Alanine-tert-butyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MW: 181.66 g/mol
Melting Pt: 168…175 °C
Flash Pt: 78.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035524
CAS Number: 13404-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR