L-Alanine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
A318333-1000G
A318333-100G
A318333-500G
A318333-5G
103810-102EA
31
USD
103810-102
76838-956
77114-696
77114-698
77571-896
L-Alanine 98%
L-Alanine
Precaución: for Research Use Only
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Melting Pt: 314.5 °C Flash Pt: 82.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064410 CAS Number: 56-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...