L-Alanine ethyl ester hydrochloride 98%

Supplier: Ambeed
Warning

A225538-25G A225538-100G A225538-500G A225538-5G
103850-084EA 17 USD
103850-084 77088-786 77275-608 77275-610 77303-744
L-Alanine ethyl ester hydrochloride 98%
L-Alanine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
Melting Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00063662
CAS Number: 1115-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR