Ethyl-1-aminocyclopropanecarboxylate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Acpc-OEt.HCl, H-Acpc-OEt·HCl
A168087-1G
A168087-25G
A168087-5G
A168087-100G
A168087-10G
103806-246EA
5
USD
103806-246
103844-504
77081-544
77081-546
77081-548
77096-418
77096-420
Ethyl-1-aminocyclopropanecarboxylate hydrochloride 95%
Ethyl-1-aminocyclopropanecarboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₂·HCl MW: 165.6 g/mol Melting Pt: 114…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190747 CAS Number: 42303-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...