(S)-2-Aminobutanamide hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Abu-NH2 · HCl, (2S)-2-Aminobutanamide hydrochloride, (S)-(+)-2-Aminobutanamide hydrochloride, H-Abu-NH2 HCl

A231365-1000G A231365-100G A231365-25G A231365-500G A231365-5G
103850-524EA 6 USD
103850-524 76874-254 76874-256 76874-258 76874-260 77303-912
(S)-2-Aminobutanamide hydrochloride 98%
(S)-2-Aminobutanamide hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁ClN₂O
MW: 138.60 g/mol
Melting Pt: 259…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00136565
CAS Number: 7682-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR