5-Aminovaleric acid 98%

Supplier: Ambeed
Warning

Synonyms: H-5-Ava-OH, 5-Aminopentanoic acid

A362530-25G A362530-5G A362530-100G A362530-10G A362530-1G
103826-324EA 6 USD
103826-324 103837-340 76840-556 76840-558 77115-316 77115-318 77306-744
5-Aminovaleric acid 98%
5-Aminovaleric acid

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 247.5±23.0 °C (760 mm Hg)
Melting Pt: 158…161 °C
Flash Pt: 103.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008232
CAS Number: 660-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR