3-(2-Naphthyl)-L-alanine 97%

Supplier: Ambeed
Warning

A145213-10G A145213-1G A145213-25G A145213-5G A145213-100G
103803-506EA 20 USD
103803-506 103841-588 76719-484 76719-486 77316-868 77316-870 77316-872 77879-025
3-(2-Naphthyl)-L-alanine 97%
3-(2-Naphthyl)-L-alanine

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
Boiling Pt: 412.3 °C (760 mm Hg)
Melting Pt: 240…253 °C
Flash Pt: 203.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00066087
CAS Number: 58438-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR