L-1-Naphthylalanine 97%

Supplier: Ambeed
Warning

Synonyms: H-1-Nal-OH, 3-(1-Naphthyl)-L-alanine

A108169-100G A108169-10G A108169-250MG A108169-25G A108169-1G A108169-5G
103797-306EA 16 USD
103797-306 103823-084 76717-928 76717-930 76717-932 76717-934 77309-926 77309-928
L-1-Naphthylalanine 97%
L-1-Naphthylalanine

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MW: 215.252 g/mol
Boiling Pt: 412.3 °C (760 mm Hg)
Melting Pt: 254…257 °C
Flash Pt: 203.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066086
CAS Number: 55516-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR