Gold trichloride 98%

Supplier: Ambeed
Warning

Synonyms: Gold(III) chloride, Gold (III) chloride

A125901-250MG A125901-1G A125901-5G
103800-736EA 85 USD
103800-736 76797-664 77298-098 77298-100
Gold trichloride 98%
Gold trichloride

Precaución: for Research Use Only

Formula: AuCl₃
MW: 303.32
Density: 4.3
Storage Temperature: Ambient
MDL Number: MFCD00014172
CAS Number: 13453-07-1
Merck Index: 13,04536

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR